Learn More
BURKLE INC Single-use sampler powders
Supplier: BURKLE INC 53935531
Simple disposable sampler for taking samples of low viscosity media If the DispoPipette is inserted into the liquid opened an all-layer sample is obtained If it is closed while it is inserted up to the sampling point and then opened a target-point sample is obtained Very easy to operate With two opening sizes (2 5/5 5 mm (0 10/0 22 in )) for different viscosities
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.