Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Disposable Protective Coveralls with Collar, 4510 Series

Catalog No. 17624101 Shop All 3M Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-624-101 X-Large
17-624-100 Medium
19-164-136 3X-Large
19-164-138 Large
17-624-102 2X-Large
19-164-137 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-624-101 Supplier 3M™ Supplier No. 7000088926
This item is not returnable. View return policy

Coveralls provide protection from certain light liquid splashes and hazardous dusts while enhancing movement and comfort.

  • Elastic waist, ankles, and wrists for convenience and freedom of movement
  • Two-way zipper with storm flap for convenience and additional protection
  • Anti-static coating on both sides helps reduce static build-up and prevent the risk of static discharge
  • Does not contain components made from natural rubber latex to help prevent reactions from users with latex sensitivities or restrictions
  • Does not contain components made from silicone to help prevent reactions from users with sensitivities or restrictions
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size X-Large
Gender Unisex
Material Laminate
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Resistance Anti-static, Dust, Liquid
Certifications/Compliance (Europe) CE, EN 1149-1:2006, EN ISO 13982-1 Type 5, EN 13034 Type 6
Latex Free Yes
Model 4510CS-BLK
For Use With (Application) Abatement, Remediation, Cleaning, Maintenance, Food Processing, Painting, Pesticide, Pharmaceutical
Quantity 25/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.