Learn More
Fisherbrand™ Disposable SMS Laboratory Coats PROMO
Catalog No. 14901000W25 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
5X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
14-901-000W25 Small
14-901-001W25 Medium
14-901-002W25 Large
14-901-003W25 X-Large
14-901-004W25 2X-Large
14-901-005W25 3X-Large
14-901-006W25 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 14-901-000W25
Lab coats that come complete with a knitted collar and cuffs. Designed to protect healthcare professionals, lab technicians and other users from fluids and cross-contamination.
Fisherbrand™ premium protective apparel is available in both lab coats and hip length jackets. Made from high quality, ultra-soft SMS fabric designed to keep you protected and comfortable.
  • Fluid-resistant material meets AAMI L2 standards
  • 42 GSM high quality, breathable SMS material
  • Anti-static coating
  • High quality front snap button closure
  • Comfortable knit collar and cuffs
  • 3-pocket design
  • Convenient side slits for easy access to pockets
Specifications
Product Type General Purpose Lab Coat
Size Small
Gender Unisex
Material SMS
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Resistance Liquids
Style Knee-length
No. of Pockets 3
Collar Type Knit
Latex Free Yes
Certifications/Compliance ANSI, AAMI PB70
Packaging Type Bagged
Autoclavable Not Autoclavable
No. per Pack 10/Pk.