Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protective Industrial Products Disposable Soft Foam Corded Ear Plugs

Catalog No. 19100426 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size: One Size
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-100-426 Size: One Size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-100-426 Supplier Protective Industrial Products Supplier No. 267HPF300C
This item is not returnable. View return policy

Features a smaller, contoured design for smaller auditory canals. Protective Industrial Products Disposable Soft Foam Corded Ear Plugs reduce the level of noise when working in a construction, manufacturing, or workshop enviroment.

  • Smaller, contoured design for smaller auditory canals
  • Polyurethane foam with a unique cellular structure allows for slower expansion recovery time, easier fitting and an improved acoustic seal
  • X Emboss at the end of plug reduces pressure at opening of ear canal for improved all day comfort and provides a grip for easy removal
  • Sealed surface prevents foreign bodies from penetrating plug

Specifications

Color Magenta
Certifications/Compliance ANSI S3.19-1974, NRR 30
Size Universal
For Use With (Application) Oil and gas, manufacturing, workshop, building and construction, and energy applications
Product Type Corded Earplug
Material Polyurethane Foam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.