Learn More
Tecan 350L Tips ANSI NF Nest
Supplier:  Tecan 30083400
Encompass_Preferred
Genuine Tecan 350 L disposable automation tips for Freedom EVO Liquid Handling Arm™ (LiHa) or Fluent Flexible Channel Arm™ (FCA). ANSI format, nested 5 stack, non-filtered, conductive. Tecan PURE purity level - tested & certified to be free of human DNA, Rnase, Dnase & PCR inhibitors. 16 tip boxes x 480 tips, 7,680 tips/sales unit. LiHa Disposable Tips have been carefully designed & validated to work specifically with Liquid Handling Arms™ on all Tecan platforms. They are produced to stringent standards of quality & reproducibility, ensuring reliable liquid handling performance. Mfg is fully automated in a controlled environment, using state-of-the-art production processes, integrated quality control procedures & premium raw materials. During production, each tip undergoes in-line optical inspection using a CCD camera & image recognition software, checking all functional parts to ensure consistent pipetting accuracy & precision.
Catalog No. NC0725393
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.