Learn More
StatLab DISSECTABLE JR.- CUTTING BOARD
SDP
Supplier:  StatLab LWC07
Encompass_Preferred
A reversible dissecting tray that contains four different size scales and a drain groove around the edge. It is constructed out of high density polyethylene and is autoclavable. Dimensions: 13 in.x11 in.x 1/2 in.
Catalog No. NC0567664
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.