Learn More
Sigma Aldrich Fine Chemicals Biosciences Mix-n-StainTM CfTM 647 Ant
Supplier: Sigma Aldrich Fine Chemicals Biosciences MX647S1001KT
This labeling kit is for labeling 50-100 mug antibody with the far red fluorescent dye Cf647. Mix-n-Stain Cf dye antibody labeling kits are a revolutionary antibody labeling technology that permits one to label antibodies with our superior Cf dyes in 30 minutes without a purification step. The labeling procedure tolerates many common buffer components and antibody stabilizers. Cf647 is far-red fluorescent dye and one of an outstanding series of Cf dyes. Cf647 is spectrally similar to Alexa fluor(R) 647 and Cy(R)5 dyes. Cf dyes are superior to Alexa fluor(R) dyes and Cy(R) dyes for antibody labeling by having combined advantages in brightness photostability specificity and novel features ideal for in vivo imaging.