Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Distance Learning: DNA Extraction
GREENER_CHOICESCI_ED

Catalog No. S08196 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S08196 7 to 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08196 Supplier Innovating Science™ Supplier No. IS1505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Discusses the history of DNA and its structure, as well as genetic inheritance; isolate DNA material from plants

  • Objectives:
    • Learn about DNA structure, history, and discovery
    • Understand the role of DNA and proteins in genetic expression
    • Use biological detergents, enzymes, and ethanol to isolate DNA from plant material
  • Includes:
    • 50 mL 7.5% SDS/ 1.5% NaCl
    • 1 pepsin (to make 25 mL 0.5% solution)
    • 10 mL ethanol, 95%
    • 2 zipper bags
    • 2 filters
    • 2 plastic tubes
    • 4 graduated pipettes
    • 2 stirrers
    • 2 plastic cups
  • Notes:
    • Plant material not included
    • Kit contains an instruction manual and enough material for two groups

Specifications

Grade 7 to 15
Age 12 to 20
Class Size 2 Lab Groups
Product Type DNA Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.