Specifications
| Includes | No Hose Hose Connection |
| For Use With (Application) | Organic Synthesis |
| Type | Distribution Adapter |
| Material | Borosilicate Glass |
| For Use With (Equipment) | Distillation Set |
| Quantity | 1 Ea. |
| Standard Taper Joint | 19/22 |
| Joint Type | Ground Glass |
| Autoclavable | Autoclavable |