Learn More
Synthware™ Distribution Adapter for Distillation Receiving Set
GREENER_CHOICESDP
Catalog No. 31501048 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint:
14/20
19/22
24/40
Includes:
Hose Connection and Angled Top
No Hose Hose Connection
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint Includes
31-501-048 19/22 Hose Connection and Angled Top
31-501-043 14/20 No Hose Hose Connection
31-501-045 19/22 No Hose Hose Connection
31-501-047 14/20 Hose Connection and Angled Top
31-501-049 24/40 Hose Connection and Angled Top
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 31-501-048 Supplier Synthware™ Supplier No. A551922
Add to Cart
Add to Cart
Designed without hose connection to be used with D10 distillation apparatus.
Specifications
Includes Hose Connection and Angled Top
For Use With (Application) Organic Synthesis
Type Distillation Adapter
Material Borosilicate Glass
For Use With (Equipment) Distillation Set
Quantity 1 Ea.
Standard Taper Joint 19/22
Joint Type Ground Glass
Autoclavable Autoclavable