Learn More
Tecan DISULFIDE HMGB1 LPS-FREE 100UG
Supplier:  Tecan REHM121
Encompass_Preferred
HMGB1, Fully reduced, LPS-free. 100 g . HMGB1, a nuclear protein released passively by necrotic cells, retained by apoptotic cells, and secreted actively by inflammatory cells. Two different activities of HMGB1 as a chemoattractant of motile cells and as inducer of cytokines have been attributed to different biochemical forms. Chemokine-HMGB1 (the form with chemoattractant activity) is completely reduced, while Cytokine-HMGB1 contains a disulfide bond between C23 and C45 and a reduced cysteine residue 106. Disulfide HMGB1 can induce cytokine and chemokine production in monocytes and other inflammatory cells. Disulfide HMGB1 is tested for the ability to stimulate cytokine production in human macrophages. The Disulfide HMGB1 we provide is the natural protein, with no tags or additional amino acids, in lyophilized form avoiding dry ice shipment. Distributed by IBL. Research Use Only.
Catalog No. 50-130-6197
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.