Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Dithiooxamide, Metal Chelating Agent, Powder, 5 g

Catalog No. 502704846
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4846 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4846 Supplier Abcam Supplier No. AB1462045G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfur analog of oxamide used for the determination of copper(II), nickel(II) and cobalt(II).

  • Molecular Weight: 120.2 g/mol
  • Form: Powder
  • Composition: 100% Dithiooxamide
  • Application: Metal chelating agent used in the determination of copper(II), nickel(II) and cobalt(II)
  • Chemical Description: Sulfur analog of oxamide

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Copper(II), Nickel(II), Cobalt(II)
Shipping Condition Ambient
Content And Storage Store at ambient temperature.
Form Powder
Quantity 5 g
For Use With (Application) Determination of copper(II), nickel(II) and cobalt(II)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.