Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dithizone, 98%

Catalog No. AAA1794714
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1794714 25 g
AAA1794706 5 g
AAA1794718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1794714 Supplier Thermo Scientific Chemicals Supplier No. A1794714
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 60-10-6 | C13H12N4S | 256.33 g/mol

Dithizone acts as an indicator for metals. It is also used as a chelating agent for heavy metals like mercury and lead. It is also used to evaluate the purification of human pancreatic islet preparations, which is used for transplantation in patients with type 1 diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dithizone acts as an indicator for metals. It is also used as a chelating agent for heavy metals like mercury and lead. It is also used to evaluate the purification of human pancreatic islet preparations, which is used for transplantation in patients with type 1 diabetes.

Solubility
Soluble in ethanol, chloroform, dimethylsulfoxide, and pyridine. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-10-6
Molecular Formula C13H12N4S
Molecular Weight (g/mol) 256.33
MDL Number MFCD00003025
InChI Key UOFGSWVZMUXXIY-UHFFFAOYSA-N
Synonym dithizone, dithizon, ditizon, carbazone, diphenylthio, usaf ek-3092, diazenecarbothioic acid, phenyl-, 2-phenylhydrazide, 1,5-diphenyl-3-mercaptoformazan, 3-formazanthiol, 1,5-diphenyl, unii-njz2cj4d6p, 1,5-diphenyl-3-thiocarbazone
PubChem CID 657262
SMILES S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1

Specifications

Melting Point ∼168°C (decomposition)
Odor Odorless
Quantity 25 g
Beilstein 748838
Merck Index 14,3377
Solubility Information Soluble in ethanol,chloroform,dimethylsulfoxide,and pyridine. Insoluble in water.
IUPAC Name 3-(phenylamino)-1-(phenylimino)thiourea
Formula Weight 256.33
Percent Purity 98%
Chemical Name or Material Dithizone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-454-1
RTECSNumber LQ9450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.