Learn More
Ditridecyl phthalate, ≥98.0% (HPLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| CAS | 119-06-2 |
| Physical Form | Neat |
| Linear Formula | C34H58O4 |
| MDL Number | MFCD00048451 |
| Synonym | 1,2-Benzenedicarboxylic acid 1,2-ditridecyl ester; Di-n-tridecyl phthalate; Phthalic acid ditridecyl ester |
| Type | Analytical Standard |
| Quantity | 50 mg |
| Formula Weight | 530.82 |
| Percent Purity | ≥98.0% (HPLC) |
| Grade | Analytical Standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.