Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dityrosine Monoclonal Antibody (7D4)
GREENER_CHOICE

Catalog No. PIMA527575
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527575 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527575 Supplier Invitrogen™ Supplier No. MA527575
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A 1:1,000 dilution of MA5-27575 was sufficient for detection of dityrosine in 1 μg of Dityrosine conjugated to BSA by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary Antibody. Specific for dityrosine modified proteins. Does not cross-react with 3,5-dibromotyrosine or bromotyrosine modified proteins.

Dityrosine may be a natural component of protein structure, a product of environmental stress, or a product of in vitro protein modification.Dityrosine is involved in diverse reactions that produce tyrosyl radicals, which in turn may couple to yield dityrosine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dityrosine
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 7D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene Alias Dityrosine; Dityrosine (DT); DT; DY
Host Species Mouse
Immunogen Synthetic Dityrosine conjugated to Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.