Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Divalproex Sodium, USP, 98.10-102%, Spectrum™ Chemical
SDP

Catalog No. 18602275 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-275 25 g Amber Glass Bottle
18-602-276 100 g Amber Glass Bottle
18-602-277 500 g Amber Glass Bottle
18-602-278 1 kg Amber Glass Bottle
18-602-279 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-602-275 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D204425GM
Add to Cart
Add to Cart

D2044, 76584-70-8, (C16H31NaO4)n

Spectrum™ Chemical Divalproex Sodium, USP is used to treat seizure disorders, certain psychiatric conditions (manic phase of bipolar disorder), and to prevent migraine headaches. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 76584-70-8
Molecular Formula C16H31NaO4
Molecular Weight (g/mol) 310.41
InChI Key MSRILKIQRXUYCT-UHFFFAOYSA-M
IUPAC Name sodium 2-propylpentanoic acid 2-propylpentanoate
SMILES [Na+].CCCC(CCC)C(O)=O.CCCC(CCC)C([O-])=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 98.10 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.