Learn More
TARGETMOL CHEMICALS INC DIVAPLON 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T27186
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Divaplon is a GABA receptor agonist (IC50 0.056 µM) that displays non-sedating anxiolytic behavioral characteristics in a rat model of anxiety. purity: 97%
Catalog No. 50-258-0639
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.