Learn More
Sigma Organic Chemistry Divinyl sulfone | 100G | 77-77-0 | MFCD00008623 | >=96%
Supplier: Sigma Organic Chemistry V3700100G
            Divinyl sulfone, 100G
About This Item:
Synonym(s): Vinyl sulfone
Linear Formula: (CH2=CH)2SO2
Molecular Weight: 118.15
CAS Number: 77-77-0
MDL Number: MFCD00008623
UNSPSC Code: 12162002
Purity: >=96%
Boiling Point: 234 C (lit.)
Melting Point: -26 C (lit.)
Density: 1.177 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.476 (lit.)
Storage: 2-8C
EINECS Number: 201-057-6
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.