Learn More
Divinyltin dichloride, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Boiling Point | 54°C to 56°C (3 mmHg) |
| Quantity | 2 g |
| UN Number | UN2788 |
| Sensitivity | Moisture sensitive |
| Formula Weight | 243.69 |
| Physical Form | Liquid |
| Chemical Name or Material | Divinyltin dichloride |
Safety and Handling
| Hazard Category | H301+H311-H330 |
| Hazard Statement | GHS H Statement H300-H310-H330 Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S. |
| EINECSNumber | 231-399-1 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.