Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dixylyl Sulfone 94.0+%, TCI America™
SDP

Catalog No. D096010G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D096010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D096010G Supplier TCI America Supplier No. D096010G

Chemical Identifiers

CAS 27043-27-2
Molecular Formula C16H18O2S
Molecular Weight (g/mol) 274.378
MDL Number MFCD00059867
InChI Key IOGPZWCEELGKFR-UHFFFAOYSA-N
PubChem CID 15741776
IUPAC Name 1-(2,3-dimethylphenyl)sulfonyl-2,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)S(=O)(=O)C2=CC=CC(=C2C)C)C

Specifications

Melting Point 117°C
Color White-Yellow
Quantity 10 g
Formula Weight 274.38
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dixylyl Sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.