Learn More
TARGETMOL CHEMICALS INC JW 67 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2288410MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. JW 67 is an inhibitor of canonical Wnt pathway signaling. JW 67 rapidly reduced active beta-catenin with a subsequent downregulation of Wnt target genes, including AXIN2, SP5, and NKD1.
Catalog No. 50-287-7692
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.