Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Velp Scientifica™ DK8 Kjeldahl Digester System
SDP

Catalog No. 07222009
Change view
Click to view available options
Voltage:
115 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
07-222-009 115 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-222-009 Supplier Velp Scientifica™ Supplier No. S30110420
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This digestion unit with an 8-position aluminum heating block offers ideal thermal homogeneity.

  • Ensures high thermal homogeneity, heating up to 450°C
  • Temperature of the block is constantly controlled by a microprocessor and is highly stable, homogeneous and precise: ±0.5°C at all selectable temperatures
  • Performs batch digestion of up to 8 samples with 250 mL, 42 mm dia. test tubes, reducing the energy consumption

Specifications

Product Type Kjeldahl Digestion Apparatus
Display Digital
Voltage 115 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.