Learn More
Velp Scientifica™ DKL8 Automatic Kjeldahl Digester System
SDP
Catalog No. 07222006
Change view
Click to view available options
Voltage:
115 V
Catalog No. Voltage
07-222-006 115 V
1 options
This item is not returnable. View return policy
Catalog No. 07-222-006 Supplier Velp Scientifica™ Supplier No. S30110200
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
This digestion unit with an 8-position aluminum heating block offers ideal thermal stability and homogeneity.
  • Automatic and immediately ready to use
  • Consist of an aluminum heating block, a lift for automatic sample handling, suction hood, test tubes, test tube rack and drip tray
  • Fast aluminum heating block is capable of reaching 420°C in just 20 min. with best-in-class temperature stability
  • Choice of premium materials and the powerful microprocessor guarantee precise block temperature with an accuracy of ±0.5°C
Specifications
Product Type Automated Digestion System with Lift
Display Graphic LCD
Voltage 115 V