Learn More
Thermo Scientific Chemicals DL-1,4-Dithiothreitol, 99%, for biochemistry
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
- 1,4-Dithiothreitol (DTT or Cleland's reagent) reduces disulfide bonds in proteins, leading to loss of native conformation
- DTT is not stable in solution and should be prepared in small aliquots prior to use; the use of EDTA in the solution can increase the half-life of DTT
- Typically used at a final concentration of 0.1M for protein stabilization; recommended to create a 1M stock
Applications
- Protects thiol groups from oxidation and acts as a chemical chelator in solution; maintains proteins in the denatured state
- Common applications include SDS-page electrophoresis and protein isolation
Specifications
Specifications
| Absorbance | (1M soln. in water), (1 M soln. in water) at 0nm, 0.035 max. at 500nm |
| Melting Point | 39.0°C to 44.0°C |
| Color | White |
| Boiling Point | 125.0°C to 130.0°C (2.0 mmHg) |
| Flash Point | >110°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (Iodimetry) |
| Packaging | Glass bottle |
| Molecular Formula | C4H10O2S2 |
| Linear Formula | HSCH2CH(OH)CH(OH)CH2SH |
| Show More |
Safety and Handling
| Hazard Category | Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2) |
| Hazard Statement | Causes serious eye irritation, Harmful if swallowed, May cause respiratory irritation, May form combustible dust concentrations in air, Causes skin irritation |
| EINECSNumber | 222-468-7 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.