Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water

Catalog No. AC426382500
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC426382500 250 mL
AC426380100 10 mL
AC426380500 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC426382500 Supplier Thermo Scientific Chemicals Supplier No. 426382500
Add to Cart
This item is not returnable. View return policy

CAS: 3483-12-3 | C4H10O2S2 | 154.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For Biochemistry
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-1, 4-Dithiothreitol
Color Colorless
Name Note 1M Solution in water, For molecular biology
CAS 3483-12-3, 7732-18-5
Molecular Formula C4H10O2S2
MDL Number MFCD00004877
Concentration 0.90 to 1.10M
Density 1.0400g/mL
Merck Index 15, 3419
Synonym dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
SMILES C(C(C(CS)O)O)S
IUPAC Name (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
Molecular Weight (g/mol) 154.24
PubChem CID 446094
ChEBI CHEBI:42170
Formula Weight 154.24
Grade Biochemical
Packaging Glass bottle
Specific Gravity 1.04
Quantity 250 mL
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.