Learn More
Sigma Organic Chemistry DL-2-Amino-1 1 1-triflu | 1G | 1582-18-9 | MFCD16875525
Supplier: Sigma Organic Chemistry MNO0001861G

DL-2-Amino-1 1 1-triflu , 1G
About this item:
CAS #: 1582-18-9
MDL #: MFCD16875525
Chemical Formula: C5H11ClF3N
Molecular Weight: 177.6
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.