Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Amino-n-octanoic Acid 98.0+%, TCI America™
SDP

Catalog No. A038225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A038225G 25 g
A03825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A038225G Supplier TCI America Supplier No. A038225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 644-90-6
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.229
MDL Number MFCD00008102
InChI Key AKVBCGQVQXPRLD-UHFFFAOYSA-N
Synonym dl-2-aminocaprylic acid, dl-2-aminooctanoic acid, 2-aminocaprylic acid, octanoic acid, 2-amino, 2-amino-dl-caprylic acid, +--2-aminooctanoic acid, dl-alpha-aminocaprylic acid, 2-amino-octanoic acid, +/--2-aminooctanoic acid, caprylic acid, alpha-amino
PubChem CID 69522
ChEBI CHEBI:75145
IUPAC Name 2-aminooctanoic acid
SMILES CCCCCCC(C(=O)O)N

Specifications

Color White
Quantity 25 g
Formula Weight 159.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-2-Amino-n-octanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.