Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Aminoadipic acid, 98%

Catalog No. AAL1271703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1271703 1 g
AAL1271706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1271703 Supplier Thermo Scientific Chemicals Supplier No. L1271703
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 542-32-5 | C6H11NO4 | 161.16 g/mol

DL-2-Aminoadipic acid is a labeled amino acid which is used in chemical research and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-2-Aminoadipic acid is a labeled amino acid which is used in chemical research and as pharmaceutical intermediate.

Solubility
Soluble in water (2.2 mg/ml at 20°C), 1 M HCl (50 mg/ml), ethanol (slightly), ether (slightly), and 80% formic acid (25 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. Keep in a dry place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-32-5
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.16
MDL Number MFCD00063119,MFCD00063118
InChI Key OYIFNHCXNCRBQI-UHFFFAOYNA-N
Synonym dl-2-aminoadipic acid, aminoadipic acid, alpha-aminoadipic acid, 2-aminoadipic acid, dl-alpha-aminoadipic acid, 2-aminoadipate, alpha-aminoadipate, hexanedioic acid, 2-amino, 2-amino-hexanedioic acid, dl-2-aminohexanedioic acid
PubChem CID 469
ChEBI CHEBI:37024
IUPAC Name 2-aminohexanedioic acid
SMILES NC(CCCC(O)=O)C(O)=O

Specifications

Melting Point 196°C to 198°C
Quantity 1 g
Beilstein 1773077
Merck Index 14,416
Solubility Information Soluble in water (2.2mg/ml at 20°C),1 M HCl (50mg/ml),ethanol (slightly),ether (slightly),and 80% formic acid (25mg/ml).
Formula Weight 161.16
Percent Purity 98%
Chemical Name or Material DL-2-Aminoadipic acid
EINECSNumber 208-809-2
RTECSNumber MO1852000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.