Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-3-(α-Acetonyl-4'-chlorobenzyl)-4-hydroxycoumarin 98%, Thermo Scientific™

Catalog No. AC183050010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC183050010 1 g
AC183050050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC183050010 Supplier Thermo Scientific Chemicals Supplier No. 183050010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-82-3
Molecular Formula C19H15ClO4
Molecular Weight (g/mol) 342.78
MDL Number MFCD00012094
InChI Key HENZOLWOIZODCT-UHFFFAOYNA-N
Synonym coumachlor, ratilan, tomorin, cumachlor, coumachlore, cumachloor, kumachlor, geigy rodenticide exp. 332, kumachlor czech, cumachloor dutch
PubChem CID 54682651
ChEBI CHEBI:80741
IUPAC Name 3-[1-(4-chlorophenyl)-3-oxobutyl]-2-hydroxy-4H-chromen-4-one
SMILES CC(=O)CC(C1=CC=C(Cl)C=C1)C1=C(O)OC2=CC=CC=C2C1=O

Specifications

Quantity 1 g
Beilstein 17, III, 6796
Merck Index 15, 2544
Formula Weight 342.77
Chemical Name or Material DL-3-(α-Acetonyl-4'-chlorobenzyl)-4-hydroxycoumarin, 98%
EINECSNumber 201-378-1
RTECSNumber GN4830000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.