Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dl-Alpha-Tocopherol Methoxypolyethylene Glycol Succinate Solution

Catalog No. 733857500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
50 mL
100 mL
500 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-385-7500ML 500 mL
73-385-710ML 10 mL
73-385-750ML 50 mL
73-385-7100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 73-385-7500ML Supplier Sigma Aldrich Supplier No. 733857500ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product TPGS-750-M is a lead surfactant for many transition metal-catalyzed cross-couplings and has been enhanced for catalytic efficiency. Find details here.Second generation amphiphile which forms micelles upon dissolution in water allowing a variety of cross-coupling reactions to take place in water instead of organic solvent.Included among the possible reactions are:Olefin metathesisPd-catalyzed cross coupling reactions including Heck, Suzuki-Miyaura, Sonogashira and Buchwald-Hartwig reactionsC-H activation reactionsSecond generation amphiphile for cross-coupling reactions in waterWatch Professor Lipshutz talk about TPGS-750-M in this webinar:From milligrams to kilograms: synthetic chemistry following nature's lead

Specifications

CAS 1309573-60-1
Density 1.002 g/mL at 25°C
Refractive Index n20/D 1.336
Linear Formula (C2H4O)nC34H56O5
Quantity 500 mL
Synonym TPGS-750-M
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.