Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-α-Amino-n-butyric Acid, MP Biomedicals

Catalog No. ICN10043090 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10043090 500 g
ICN10043080 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10043090 Supplier MP Biomedicals, Inc Supplier No. 0210043090
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56-12-2
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00008093
InChI Key BTCSSZJGUNDROE-UHFFFAOYSA-N
Synonym 4-aminobutyric acid, gaba, gamma-aminobutyric acid, piperidic acid, piperidinic acid, aminalon, gaballon, gammalon, mielogen, butanoic acid, 4-amino
PubChem CID 119
ChEBI CHEBI:16865
IUPAC Name 4-aminobutanoic acid
SMILES C(CC(=O)O)CN

Specifications

Quantity 500 g
Formula Weight 103.12
Chemical Name or Material DL-Alpha-Amino-n-Butyric Acid
EINECSNumber 220-616-5
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.