accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOHO

CAS RN 56-12-2

IUPAC Name: 4-aminobutanoic acid
Synonyms: gamma(amino)-butyric acid immu-G 4Abu w-aminobutyrate gamma-amino-N-butyric acid acide amino-4- butyrique GABA gaba, lithium reanal epa pesticide chemical code 030802
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: BTCSSZJGUNDROE-UHFFFAOYSA-N
SMILES: NCCCC(O)=O
Molecular Weight (g/mol) 
  • (9)
  • (9)
Quantity 
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (5)
Percent Purity 
  • (1)
  • (5)
  • (3)
  • (3)
  • (5)
Grade 
  • (5)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (5)
  • (1)
  • (2)
  • (5)
  • (3)
  • (8)
  • (1)
  • (1)
  • (1)
  • (9)
  • (9)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (5)
  • (1)
  • (5)
  • (3)
  • (3)
  • (5)
  • (5)
  • (2)