Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Cystine

Catalog No. AAJ6356422
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6356422 100 g
AAJ6356406 5 g
AAJ6356414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6356422 Supplier Thermo Scientific Chemicals Supplier No. J6356422
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 923-32-0 | C6H12N2O4S2 | 240.292 g/mol

DL-cystine is used in the preparation of sulfur-containing dimeric and monomeric surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-cystine is used in the preparation of sulfur-containing dimeric and monomeric surfactants.

Solubility
Soluble in dilute acid and alkali solution, highly soluble in water, not soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 923-32-0
Molecular Formula C6H12N2O4S2
Molecular Weight (g/mol) 240.292
MDL Number MFCD00084652
InChI Key LEVWYRKDKASIDU-UHFFFAOYSA-N
Synonym dl-cystine, cystine, d-cystine, 3,3'-disulfanediylbis 2-aminopropanoic acid, dicysteine, cystin, h-dl-cys-oh 2, .beta.,.beta.'-dithiodialanine, cystine, dl, cystine, l
PubChem CID 595
ChEBI CHEBI:17376
IUPAC Name 2-amino-3-[(2-amino-2-carboxyethyl)disulfanyl]propanoic acid
SMILES C(C(C(=O)O)N)SSCC(C(=O)O)N

Specifications

Melting Point 227°C (decomposition)
Color White
Beilstein 1728095
Merck Index 14,2782
Solubility Information Soluble in dilute acid and alkali solution,highly soluble in water,not soluble in alcohol.
Quantity 100 g
Formula Weight 240.3
Physical Form Powder
Chemical Name or Material DL-Cystine
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 213-094-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.