Learn More
Sigma Aldrich Fine Chemicals Biosciences DL-Dithiothreitol, 3483-12-3, MFCD00004877, 10 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D977910G
Encompass
Linear Formula: HSCH2CH(OH)CH(OH)CH2SH, Molecular Weight: 154.25, for molecular biology, ≥98% (HPLC), ≥99% (titration), Synonym: threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT. An excellent reagent for maintaining SH groups in reduced state; quantitatively reduces disulfides.
Catalog No. NC0173096
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.