Learn More
Medchemexpress LLC DL-homocysteine thiolactone-d4 hydrochloride | 1219805-31-8 | MFCD00055960 | 99.8% | 157.66 g/mol | C4H4D4ClNOS | 10 MG
Supplier: Medchemexpress LLC HY101404S10MG
DL-Homocysteine thiolactone-d4 hydrochloride is a deuterium-labeled analytical standard of DL-homocysteine thiolactone supplied as the hydrochloride salt. It is designed for research and analytical applications, particularly as an internal standard in mass spectrometry, isotope-dilution assays, and isotopic tracing studies.
- Deuterium-labeled (d4) analogue for isotope-dilution assays.
- Hydrochloride salt with high chemical purity (99.83%).
- Suitable as an internal standard for LC-MS/MS and GC-MS methods.
- Solid form for easy handling and storage.
- Available in small analytical quantities for method development and QC.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.