Learn More
Medchemexpress LLC DL-homocysteine thiolactone hydrochloride | 6038-19-3 | MFCD00012724 | 100.0% | 153.63 g/mol | C4H8ClNOS | 25 G
Supplier: Medchemexpress LLC HY10140425G
DL-homocysteine thiolactone hydrochloride is a cyclic amino acid derivative supplied as the hydrochloride salt for use as a research reagent. It exhibits root-growth inhibitory activity at low micromolar concentrations and is used as a biochemical intermediate and tool compound in plant and biochemical studies. For research use only.
- High purity, 99.97% as supplied.
- Solid powder, suitable for analytical and preparative use.
- Molecular weight 153.63 g/mol; formula C4H8ClNOS.
- Exhibits root-growth inhibitory activity at low micromolar levels.
- Available in multiple gram-scale sizes, including 25 g.
- Certificate of analysis (COA) and safety data sheet (SDS) available.