Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Lactic acid, 80-85% aq. soln.

Catalog No. AAL142590B
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL142590B 1000 g
AAL1425930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL142590B Supplier Thermo Scientific Chemicals Supplier No. L142590B
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 50-21-5 | C3H6O3 | 90.08 g/mol

DL-Lactic acid is used to produce serial products or widely used in food, vintage, beverage, drugs, polymerization, textile, leather, tobacco, feed, plastic chemicals, pesticide, polymer solution and other industry. Lactic acid is also used as an acidifying agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water and in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-21-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

Boiling Point 122°C (15 mmHg)
Density 1.209
Quantity 1000 g
Flash Point >110°C (230°F)
Refractive Index 1.423
Beilstein 1209341
Merck Index 145336
Solubility Information Soluble in water and in ethanol.
Formula Weight 90.08
Concentration or Composition (by Analyte or Components) 80 to 85% aq. Soln.
Chemical Name or Material DL-Lactic acid
Show More Show Less
Hazard Category H315-H318
Hazard Statement GHS H Statement
H318-H315
Causes serious eye damage.
Causes skin irritation.
Precautionary Statement P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390
EINECSNumber 200-018-0
RTECSNumber OD2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.