Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam DL-Lactic Acid Lithium Salt, Racemic Mixture, >99% Purity, Solid, 50 g

Catalog No. 502704778
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4778 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4778 Supplier Abcam Supplier No. AB14626550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity lithium salt of racemic DL-lactic acid.

  • Molecular Weight: 96.01 g/mol
  • Purity: >99%
  • Composition: 100% Lithium(I) 2.hydroxypropanoate
  • Form: Solid
  • Description: Racemic mixture of DL isomers of the lithium salt of lactic acid.
  • CAS Number: 867-55-0
  • Storage Conditions: Store at ambient temperature

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Lactic Acid
Content And Storage Store at ambient temperature.
Form Solid
Shipping Condition Ambient
Quantity 50 g
For Use With (Application) Sample Prep and Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.