Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Lactic Acid, Racemic, Spectrum™ Chemical
SDP

Catalog No. 18620982 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-982 500 mL
18-620-983 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-982 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L2016500ML
Add to Cart
Add to Cart

Synonym: 2-Hydroxypropanoic Acid, 2-Hydroxypropionic Acid, Milk Acid

DL-Lactic Acid, also known as milk acid, is used in various biochemical processes. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 50-21-5

Specifications

Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.