Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Leucic Acid, Spectrum™ Chemical
SDP

Synonym: 2-Hydroxy-4-Methylpentanoic Acid, 2-Hydroxy-4-Methylvaleric Acid, 2-Hydroxyisocaproic Acid

Supplier:  Spectrum Chemical Manufacturing Corporation L161025GM

Quantity 25 g

 View more versions of this product

Catalog No. 18-620-974


Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart

Description

Description

Specifications

Chemical Identifiers

10303-64-7
132.16 g/mol

Specifications

10303-64-7
98% Min.
25 g
132.16 g/mol
Amber Bottle
DL-Leucic Acid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $500 in Chemicals with Purchase
Spend $250 or more on Spectrum Chemical chemicals and get up to $500 to spend on additional chemicals.

Adding...

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.