Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Leucic Acid, Spectrum™ Chemical
SDP

Synonym: 2-Hydroxy-4-Methylpentanoic Acid, 2-Hydroxy-4-Methylvaleric Acid, 2-Hydroxyisocaproic Acid

$140.00 - $386.00

Chemical Identifiers

CAS 10303-64-7
Molecular Weight (g/mol) 132.16 g/mol
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
18-620-974
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
L161025GM
25 g
Each for $140.00
Only null left
Add to Cart
 
18-620-975
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
L1610100GM
100 g
Each for $386.00
Only null left
Add to Cart
 
Description

Description

Specifications

Chemical Identifiers

10303-64-7
132.16 g/mol

Specifications

10303-64-7
98% Min.
DL-Leucic Acid
132.16 g/mol
Amber Bottle
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.