Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Lysine Monohydrochloride 98.0+%, TCI America™
SDP

Catalog No. L0070100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0070100G 100 g
L007025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L0070100G Supplier TCI America Supplier No. L0070100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 70-53-1
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00064563
InChI Key BVHLGVCQOALMSV-UHFFFAOYSA-N
Synonym dl-lysine monohydrochloride, 2,6-diaminohexanoic acid hydrochloride, dl-lysine hydrochloride, lysine monohydrochloride, dl-lysine mono hydrochloride, lysine, hydrochloride 1:1, lysine, monohydrochloride, dl-2,6-diaminohexanoic acid monohydrochloride, lysine, dl, dl-lysinemonohydrochloride
PubChem CID 12047
IUPAC Name 2,6-diaminohexanoic acid hydrochloride
SMILES Cl.NCCCCC(N)C(O)=O

Specifications

Color White-Yellow
Quantity 100 g
Formula Weight 182.65
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Lysine Monohydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.