Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals DL-Methionine, 99+%

Catalog No. AC125650025
Change view
Click to view available options
Quantity:
2.5 g
100 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125650025 2.5 g Glass Bottle
AC125651000 100 g Plastic Bottle
AC125652500 250 g Plastic Bottle
AC125650010 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC125650025 Supplier Thermo Scientific Chemicals Supplier No. 125650025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 59-51-8 | C5H11NO2S | 149.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-51-8
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063096
InChI Key FFEARJCKVFRZRR-UHFFFAOYNA-N
Synonym dl-methionine, methionine, racemethionine, acimetion, banthionine, cynaron, lobamine, meonine, mertionin, metione
PubChem CID 876
ChEBI CHEBI:16811
SMILES CSCCC(N)C(O)=O

Specifications

Ammonium 1000ppm max.
pH 5.4 to 6.1 (2% w/v aq. soln.)
Color White
Chloride 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Loss on Drying 0.5% max. (1 g, 105°C)
Other Amino Acids 0.2% max. (TLC)
Packaging Glass Bottle
Linear Formula CH3SCH2CH2CH(NH2)CO2H
Beilstein 04, II, 938
Sulfate 200ppm max.
Merck Index 15, 6047
Transmittance 95% min. (c=10, 2N HCl, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Density 1.34g/mL
IUPAC Name 2-amino-4-(methylsulfanyl)butanoic acid
Quantity 2.5 g
Formula Weight 149.21
Percent Purity 99.0 to 101.0% (on dried substance)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material DL-Methionine, 99+%
Show More Show Less
EINECSNumber 200-432-1
RTECSNumber PD0456000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.