Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals DL-Methionine sulfoxide, 98+%

Catalog No. AAA1808106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1808106 5 g
AAA1808114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1808106 Supplier Thermo Scientific Chemicals Supplier No. A1808106
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 62697-73-8 | C5H11NO3S | 165.21 g/mol

DL-Methionine sulfoxide is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Methionine sulfoxide is used as pharmaceutical intermediates.

Solubility
Soluble in water and Hcl.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62697-73-8
Molecular Formula C5H11NO3S
Molecular Weight (g/mol) 165.21
MDL Number MFCD00002620
InChI Key QEFRNWWLZKMPFJ-UHFFFAOYNA-N
Synonym dl-methionine sulfoxide, methionine sulfoxide, 2-amino-4-methylsulfinyl butanoic acid, dl-methioninesulfoxide, methionine, s-oxide, uethionine, s-oxide, butanoic acid, 2-amino-4-methylsulfinyl, 2-amino-4-methylsulfinyl butyric acid, 2-amino-4-methanesulfinylbutanoic acid, butyric acid, 2-amino-4-methylsulfinyl
PubChem CID 847
ChEBI CHEBI:49033
SMILES CS(=O)CCC(N)C(O)=O

Specifications

Melting Point ∼230°C (decomposition)
Solubility Information Soluble in water and Hcl.
IUPAC Name 2-amino-4-methylsulfinylbutanoic acid
Quantity 5 g
Formula Weight 165.21
Percent Purity ≥98%
Chemical Name or Material DL-Methionine sulfoxide
EINECSNumber 263-700-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.