Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Pantolactone 95.0+%, TCI America™
SDP

Catalog No. P0010100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0010100G 100 g
P001025G 25 g
P0010500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0010100G Supplier TCI America Supplier No. P0010100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 79-50-5
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00064333
InChI Key SERHXTVXHNVDKA-UHFFFAOYSA-N
Synonym pantolactone, dl-pantolactone, 3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, pantoyl lactone, pantoic lactone, dl-pantoyl lactone, d---pantolactone, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl, dihydro-3-hydroxy-4,4-dimethyl-2 3h-furanone, pantolyl lactone
PubChem CID 989
IUPAC Name 3-hydroxy-4,4-dimethyloxolan-2-one
SMILES CC1(COC(=O)C1O)C

Specifications

Melting Point 78°C
Color White
Boiling Point 121°C
Quantity 100 g
Formula Weight 130.14
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material DL-Pantolactone
EINECSNumber (9)-0997
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.