Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Pipecolic Acid 98.0+%, TCI America™
SDP

Catalog No. P0442250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0442250G 250 g
P044225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0442250G Supplier TCI America Supplier No. P0442250G

Chemical Identifiers

CAS 535-75-1
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00064347,MFCD00005981
InChI Key HXEACLLIILLPRG-UHFFFAOYNA-N
Synonym dl-pipecolinic acid, pipecolic acid, pipecolinic acid, 2-piperidinecarboxylic acid, homoproline, pipecolate, dl-pipecolic acid, dihydrobaikiane, piperolinic acid, hexahydropicolinic acid
PubChem CID 849
ChEBI CHEBI:17964
IUPAC Name piperidine-2-carboxylic acid
SMILES OC(=O)C1CCCCN1

Specifications

Color White
Quantity 250 g
Formula Weight 129.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Pipecolic Acid
RTECSNumber TK6021000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.