Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Proline, 99%, Thermo Scientific Chemicals

Catalog No. AAA1313714
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1313714 25 g
AAA1313706 5 g
AAA1313722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1313714 Supplier Thermo Scientific Chemicals Supplier No. A1313714
Add to Cart

DL-Proline is used as an agonist of glycine receptors. It acts as a catalyst for aldol condensation. It is an essential component of collagen. It plays a significant role in the immune system and strengthening of heart muscles. It is an osmoprotectant and therefore is used in many pharmaceutical, biotechnological applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Proline is used as an agonist of glycine receptors. It acts as a catalyst for aldol condensation. It is an essential component of collagen. It plays a significant role in the immune system and strengthening of heart muscles. It is an osmoprotectant and therefore is used in many pharmaceutical, biotechnological applications.

Solubility
Soluble in water and absolute alcohol. Insoluble in ether, butanol and isopropanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-36-9
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00005250
InChI Key ONIBWKKTOPOVIA-UHFFFAOYSA-N
Synonym dl-proline, proline, h-dl-pro-oh, proline, dl, 2-pyrrolidine carboxylic acid, hpro, l-proline, homopolymer, +/--pyrrolidine-2-carboxylic acid, prolin, chembl72275
PubChem CID 614
ChEBI CHEBI:26271
IUPAC Name pyrrolidine-2-carboxylic acid
SMILES C1CC(NC1)C(=O)O

Specifications

Melting Point ∼209°C (decomposition)
Beilstein 80809
Merck Index 14,7780
Solubility Information Soluble in water and absolute alcohol. Insoluble in ether,butanol and isopropanol.
Quantity 25 g
Formula Weight 115.13
Percent Purity 99%
Chemical Name or Material DL-Proline
EINECSNumber 210-189-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.