Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals DL-Selenomethionine, 99+%

Catalog No. AC240770010
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC240770010 1 g Glass bottle
AC240775000 500 mg Glass bottle
AC240770025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC240770010 Supplier Thermo Scientific Chemicals Supplier No. 240770010
Add to Cart

CAS: 1464-42-2 | C5H11NO2Se | 196.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1464-42-2
Molecular Formula C5H11NO2Se
Molecular Weight (g/mol) 196.12
MDL Number MFCD00063089
InChI Key RJFAYQIBOAGBLC-UHFFFAOYNA-N
Synonym selenomethionine, dl-selenomethionine, seleno-dl-methionine, +--selenomethionine, butanoic acid, 2-amino-4-methylseleno, selenium methionine, methionine, seleno, selenomethionine van, 2-amino-4-methylselanyl butanoic acid, seleno-d,l-methionine
PubChem CID 15103
ChEBI CHEBI:27585
IUPAC Name 2-amino-4-methylselanylbutanoic acid
SMILES C[Se]CCC(N)C(O)=O

Specifications

Melting Point 265.0°C
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Odor Faint Smelling
Packaging Glass bottle
Linear Formula CH3SeCH2CH2CH(NH2)COOH
Quantity 1 g
Merck Index 15, 8578
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, insoluble in benzene, diethyl ether
Formula Weight 196.11
Percent Purity 99+%
Physical Form Glistening Powder
Chemical Name or Material DL-Selenomethionine, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 215-977-
RTECSNumber ES7110000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.