Learn More
Bachem DL-Thiorphan
SDP
Supplier:  Bachem 4010438.0025
Encompass_Preferred
DL-Thiorphan , ((RS)-2-Benzyl-3-mercaptopropionyl)-Gly-OH, 76721-89-6, C12H15NO3S, Mr 253.32, 25mg. (DL-3-Mercapto-2-benzylpropanoyl)-Gly-OH. Potent and specific inhibitor of neprilysin with antinociceptive activity. Thiorphan is cleaved by peptidylglycine a-amidating monooxygenase yielding 2-benzyl-3-mercaptopropionamide.
Catalog No. 50-260-527
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.