Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals DL-Threonine, 98+%

Catalog No. AC138941000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC138941000 100 g Glass Bottle
AC138940250 25 g Glass Bottle
AC138945000 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC138941000 Supplier Thermo Scientific Chemicals Supplier No. 138941000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 80-68-2 | C4H9NO3 | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-68-2
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00063722
InChI Key AYFVYJQAPQTCCC-PWNYCUMCSA-N
Synonym dl-threonine, allo-dl-threonine, threonine, dl, dl-allothreonine, dl-2-amino-3-hydroxybutanoic acid, threonine l, h-dl-thr-oh, dl-allo-threonine, allothreonine, d, wln: qy1&yzvq-l
PubChem CID 205
ChEBI CHEBI:38263
SMILES C[C@@H](O)[C@@H](N)C(O)=O

Specifications

Arsenic (As) 2ppm max.
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Assay Percent Range 98+%
Loss on Drying 0.2% max. (1 g, 105°C, 3 hrs)
Other Amino Acids (TLC) Passes Test
Packaging Glass Bottle
Linear Formula CH3CH(OH)CH(NH2)CO2H
Beilstein 04, 514
Merck Index 10, 9229
Sulfated Ash 0.1% max.
IUPAC Name 2-amino-3-hydroxybutanoic acid
Quantity 100 g
Formula Weight 119.12
Percent Purity 98+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material DL-Threonine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.